MAT047028678 Matrix Scientific

2-Chloro-4-methyl-quinolin-6-ol

  • MDL number MFCD00579691
  • Cas Number : 41957-91-9
  • Molecular Weight : 193.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/009878.html

Safety Information

MSDS http://www.matrixscientific.com/009878.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart