MAT047028713 Matrix Scientific

N-(6-Amino-4-methyl-benzothiazol-2-yl)-acetamide

  • MDL number MFCD00781834
  • Cas Number : 314033-48-2
  • Molecular Weight : 221.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/009910.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009910.html
Availability details for
Quantity
  • Availability
  • Add to Cart