MAT047028714 Matrix Scientific

N-(6-Amino-4-methoxy-benzothiazol-2-yl)-acetamide

  • MDL number MFCD00781848
  • Cas Number : 314033-54-0
  • Molecular Weight : 237.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/009911.html

Safety Information

MSDS http://www.matrixscientific.com/009911.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart