MAT047028716 Matrix Scientific

N-(6-Methyl-pyridin-2-yl)-succinamic acid

  • MDL number MFCD01005030
  • Cas Number : 186320-28-5
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-115
Country of Origin USA
Product Link http://www.matrixscientific.com/009913.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009913.html
Availability details for
Quantity
  • Availability
  • Add to Cart