MAT047028717 Matrix Scientific

1-Iodo-2,2,3,3-tetrafluoropropane

  • MDL number MFCD00019033
  • Cas Number : 679-87-8
  • Molecular Weight : 241.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 2.036
Storage temp. Refrigerate
Country of Origin USA
Boiling point 95-97°
Product Link http://www.matrixscientific.com/001937.html

Safety Information

MSDS http://www.matrixscientific.com/001937.html
pictograms Irritant/Light Sensitive/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart