MAT047028721 Matrix Scientific

5-Benzooxazol-2-yl-2-methoxy-phenylamine

  • MDL number MFCD00298038
  • Cas Number : 378222-48-1
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/009917.html

Safety Information

MSDS http://www.matrixscientific.com/009917.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart