MAT047028738 Matrix Scientific

5-m-Tolyloxymethyl-[1,3,4]thiadiazol-2-ylamine

  • MDL number MFCD01028941
  • Cas Number : 84138-76-1
  • Molecular Weight : 221.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202
Country of Origin USA
Product Link http://www.matrixscientific.com/009932.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009932.html
Availability details for
Quantity
  • Availability
  • Add to Cart