MAT047028749 Matrix Scientific

5-(3,4-Dimethoxy-phenyl)-[1,3,4]thiadiazol-2-yl-amine

  • MDL number MFCD00981427
  • Cas Number : 5427-87-2
  • Molecular Weight : 237.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/009942.html

Safety Information

MSDS http://www.matrixscientific.com/009942.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart