MAT047028772 Matrix Scientific

7-Phenyl-tricyclo[4.1.0.0*2,7*]heptane-1-carboxylic acid

  • MDL number MFCD08276217
  • Cas Number : 129576-92-7
  • Molecular Weight : 214.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168
Country of Origin USA
Product Link http://www.matrixscientific.com/009963.html

Safety Information

MSDS http://www.matrixscientific.com/009963.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart