MAT047028784 Matrix Scientific

Ethyl 2-(2,4-difluorophenyl)thiazole-4-carboxylate

  • MDL number MFCD00277200
  • Cas Number : 175276-93-4
  • Molecular Weight : 269.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-52
Country of Origin USA
Product Link http://www.matrixscientific.com/001944.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001944.html
Availability details for
Quantity
  • Availability
  • Add to Cart