MAT047028824 Matrix Scientific

4-(2-Hydroxy-ethylamino)-furazan-3-carboxylic acid

  • MDL number MFCD00984735
  • Cas Number : 147194-40-9
  • Molecular Weight : 173.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010010.html

Safety Information

MSDS http://www.matrixscientific.com/010010.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart