MAT047028827 Matrix Scientific

4,5,6,7-Tetrahydro-1H-imidazo[4,5-c]pyridine-6-carboxylic acid

  • MDL number MFCD00222266
  • Cas Number : 495-77-2
  • Molecular Weight : 167.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 265
Country of Origin USA
Product Link http://www.matrixscientific.com/010013.html

Safety Information

MSDS http://www.matrixscientific.com/010013.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart