MAT047028842 Matrix Scientific

3-(5-Amino-[1,3,4]thiadiazol-2-yl)-chromen-2-one

  • MDL number MFCD01539520
  • Cas Number : 156177-02-5
  • Molecular Weight : 245.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010025.html

Safety Information

MSDS http://www.matrixscientific.com/010025.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart