MAT047028876 Matrix Scientific

[2-(3,5-Dioxo-2,3,4,5-tetrahydro-[1,2,4]triazin-6-ylamino)-acetylamino]-acetic acid

  • MDL number MFCD02293887
  • Cas Number : 333432-84-1
  • Molecular Weight : 243.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010056.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010056.html
Availability details for
Quantity
  • Availability
  • Add to Cart