MAT047028889 Matrix Scientific

5-(2-Thienyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD00139081
  • Cas Number : 10551-15-2
  • Molecular Weight : 184.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201-203
Country of Origin USA
Product Link http://www.matrixscientific.com/010068.html

Safety Information

MSDS http://www.matrixscientific.com/010068.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart