MAT047028891 Matrix Scientific

2-(4-Formyl-2-methoxy-phenoxy)-N-phenyl-acetamide

  • MDL number MFCD01156958
  • Cas Number : 31438-73-0
  • Molecular Weight : 285.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010070.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010070.html
Availability details for
Quantity
  • Availability
  • Add to Cart