MAT047028910 Matrix Scientific

(2-Pyridin-2-yl-ethyl)-(2-vinyloxy-ethyl)-amine

  • MDL number MFCD02824255
  • Cas Number : 625410-03-9
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.0285/20
Country of Origin USA
Boiling point 132-135�Torr
Product Link http://www.matrixscientific.com/010087.html

Safety Information

MSDS http://www.matrixscientific.com/010087.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart