MAT047028969 Matrix Scientific

N-(4-Thiophen-2-yl-thiazol-2-yl)-phthalamic acid

  • MDL number MFCD02048054
  • Cas Number : 325807-37-2
  • Molecular Weight : 330.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010140.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010140.html
Availability details for
Quantity
  • Availability
  • Add to Cart