MAT047028970 Matrix Scientific

7-Chloro-8-methyl-4-oxo-1,4-dihydro-quinoline-3-carboxylic acid

  • MDL number MFCD01912283
  • Cas Number : 302949-00-4
  • Molecular Weight : 237.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >280 dec.
Country of Origin USA
Product Link http://www.matrixscientific.com/010141.html

Safety Information

MSDS http://www.matrixscientific.com/010141.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart