MAT047028978 Matrix Scientific

4-(5-Chlorocarbonyl-furan-2-yl)-benzoic acid ethylester

  • MDL number MFCD02258008
  • Cas Number : 333435-04-4
  • Molecular Weight : 278.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010148.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010148.html
Availability details for
Quantity
  • Availability
  • Add to Cart