MAT047028984 Matrix Scientific

3-Chloro-2,6-difluoroanisole

  • MDL number MFCD01631331
  • Cas Number : 261762-38-3
  • Molecular Weight : 178.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/001963.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001963.html
Availability details for
Quantity
  • Availability
  • Add to Cart