MAT047028996 Matrix Scientific

1-[1-(4-Amino-furazan-3-yl)-5-methyl-1H-[1,2,3]triazol-4-yl]-ethanone

  • MDL number MFCD00746845
  • Cas Number : 313952-93-1
  • Molecular Weight : 208.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010164.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010164.html
Availability details for
Quantity
  • Availability
  • Add to Cart