MAT047029000 Matrix Scientific

5-(4-Fluoro-benzyl)-thiazol-2-ylamine

  • MDL number MFCD02214714
  • Cas Number : 315241-39-5
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-136
Country of Origin USA
Product Link http://www.matrixscientific.com/010168.html

Safety Information

MSDS http://www.matrixscientific.com/010168.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart