MAT047029023 Matrix Scientific

3-Amino-4-[(pyridin-3-ylmethyl)-amino]-benzoic acid

  • MDL number MFCD03274476
  • Cas Number : 436088-89-0
  • Molecular Weight : 243.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010189.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010189.html
Availability details for
Quantity
  • Availability
  • Add to Cart