MAT047029042 Matrix Scientific

3-Methyl-4-oxo-3,4-dihydro-phthalazine-1-carboxylic acid

  • MDL number MFCD02712615
  • Cas Number : 16015-47-7
  • Molecular Weight : 204.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236�ec)
Country of Origin USA
Product Link http://www.matrixscientific.com/010206.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010206.html
Availability details for
Quantity
  • Availability
  • Add to Cart