MAT047029056 Matrix Scientific

6,11-Dihydro-5H-dibenzo[b,e]azepine

  • MDL number MFCD03723999
  • Cas Number : 449-55-8
  • Molecular Weight : 195.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-130
Country of Origin USA
Product Link http://www.matrixscientific.com/010219.html

Safety Information

MSDS http://www.matrixscientific.com/010219.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart