MAT047029077 Matrix Scientific

(6-Dimethylsulfamoyl-3-oxo-1,3-dihydro-isobenzofuran-1-yl)-acetic acid

  • MDL number MFCD00423679
  • Cas Number : 144402-61-9
  • Molecular Weight : 299.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/009853.html

Safety Information

MSDS http://www.matrixscientific.com/009853.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart