MAT047029080 Matrix Scientific

4-(1-Amino-2-phenyl-ethyl)-hepta-1,6-dien-4-ol

  • MDL number MFCD02020766
  • Cas Number : 46759-27-7
  • Molecular Weight : 231.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010241.html

Safety Information

MSDS http://www.matrixscientific.com/010241.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart