MAT047029084 Matrix Scientific

8-Methyl-thieno[2,3-b]quinoline-2-carboxylic acid

  • MDL number MFCD02228298
  • Cas Number : 333312-05-3
  • Molecular Weight : 243.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010244.html

Safety Information

MSDS http://www.matrixscientific.com/010244.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart