MAT047029129 Matrix Scientific

3-Amino-7-methoxy-thieno[2,3-b]quinoline-2-carboxylic acid

  • MDL number MFCD02590326
  • Cas Number : 462068-01-5
  • Molecular Weight : 274.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010285.html

Safety Information

MSDS http://www.matrixscientific.com/010285.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart