MAT047029132 Matrix Scientific

3-(4-Fluoro-phenylamino)-3H-isobenzofuran-1-one

  • MDL number MFCD01417302
  • Cas Number : 309926-32-7
  • Molecular Weight : 243.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-189
Country of Origin USA
Product Link http://www.matrixscientific.com/010288.html

Safety Information

MSDS http://www.matrixscientific.com/010288.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart