MAT047029134 Matrix Scientific

Benzyl-(2-methoxy-benzyl)-amine

  • MDL number MFCD00478083
  • Cas Number : 69875-89-4
  • Molecular Weight : 227.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.0678/25
Country of Origin USA
Boiling point 200� Torr
Product Link http://www.matrixscientific.com/010290.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010290.html
Availability details for
Quantity
  • Availability
  • Add to Cart