MAT047029155 Matrix Scientific

5-Amino-2-(5-methyl-benzooxazol-2-yl)-phenol

  • MDL number MFCD00440139
  • Cas Number : 293737-91-4
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010309.html

Safety Information

MSDS http://www.matrixscientific.com/010309.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart