MAT047029156 Matrix Scientific

5-Amino-2-(5-chloro-benzooxazol-2-yl)-phenol

  • MDL number MFCD00440136
  • Cas Number : 22105-55-1
  • Molecular Weight : 260.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 272-273
Country of Origin USA
Product Link http://www.matrixscientific.com/010310.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010310.html
Availability details for
Quantity
  • Availability
  • Add to Cart