MAT047029159 Matrix Scientific

2-(4-Methoxy-phenyl)-benzooxazol-5-ylamine

  • MDL number MFCD00991831
  • Cas Number : 54995-53-8
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010313.html

Safety Information

MSDS http://www.matrixscientific.com/010313.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart