MAT047029161 Matrix Scientific

4-Chloro-2-(2,5-dimethyl-pyrrol-1-yl)-benzoic acid

  • MDL number MFCD02194709
  • Cas Number : 335209-41-1
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010314.html

Safety Information

MSDS http://www.matrixscientific.com/010314.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart