MAT047029172 Matrix Scientific

4-Fluoro-2-(trifluoromethyl)benzyl alcohol

  • MDL number MFCD00061170
  • Cas Number : 220227-29-2
  • Molecular Weight : 194.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-57
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/001982.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001982.html
Availability details for
Quantity
  • Availability
  • Add to Cart