MAT047029175 Matrix Scientific

2-(3-Methoxy-phenyl)-benzooxazol-5-ylamine

  • MDL number MFCD00991832
  • Cas Number : 313527-38-7
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010326.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010326.html
Availability details for
Quantity
  • Availability
  • Add to Cart