MAT047029188 Matrix Scientific

2-[4-(2-Methoxy-phenyl)-2,2-dimethyl-tetrahydro-pyran-4-yl]-ethylamine

  • MDL number MFCD01545301
  • Cas Number : 313692-98-7
  • Molecular Weight : 263.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient/argon
Country of Origin USA
Product Link http://www.matrixscientific.com/010338.html

Safety Information

hcodes Irritant/Stored under Argon
MSDS http://www.matrixscientific.com/010338.html
Availability details for
Quantity
  • Availability
  • Add to Cart