MAT047029189 Matrix Scientific

[4-(4-Methoxy-phenyl)-2,2-dimethyl-tetrahydro-pyran-4-yl]-acetic acid

  • MDL number MFCD01544837
  • Cas Number : 337486-10-9
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010339.html

Safety Information

MSDS http://www.matrixscientific.com/010339.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart