MAT047029204 Matrix Scientific

2-p-Tolylamino-ethanol

  • MDL number MFCD00043535
  • Cas Number : 2933-74-6
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-43
Country of Origin USA
Boiling point 153-155� Torr
Product Link http://www.matrixscientific.com/010353.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010353.html
Availability details for
Quantity
  • Availability
  • Add to Cart