MAT047029205 Matrix Scientific

4-Fluoro-2-(trifluoromethyl)-phenylacetic acid

  • MDL number MFCD00061189
  • Cas Number : 195447-80-4
  • Molecular Weight : 222.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-86
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/001985.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001985.html
Availability details for
Quantity
  • Availability
  • Add to Cart