MAT047029208 Matrix Scientific

Allyl-(4-chloro-phthalazin-1-yl)-amine

  • MDL number MFCD01327243
  • Cas Number : 505071-30-7
  • Molecular Weight : 219.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010356.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010356.html
Availability details for
Quantity
  • Availability
  • Add to Cart