MAT047029214 Matrix Scientific

(4,6-Dimethyl-2-oxo-2H-quinolin-1-yl)-acetic acid

  • MDL number MFCD02926077
  • Cas Number : 370841-56-8
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010362.html

Safety Information

MSDS http://www.matrixscientific.com/010362.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart