MAT047029215 Matrix Scientific

3-(4,6-Dimethyl-2-oxo-2H-quinolin-1-yl)-propionic acid

  • MDL number MFCD02926040
  • Cas Number : 370841-34-2
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010363.html

Safety Information

MSDS http://www.matrixscientific.com/010363.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart