MAT047029220 Matrix Scientific

3-Hydroxy-2-[(5-hydroxy-pyridine-3-carbonyl)-amino]-propionic acid

  • MDL number MFCD02221583
  • Molecular Weight : 226.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010367.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010367.html
Availability details for
Quantity
  • Availability
  • Add to Cart