MAT047029222 Matrix Scientific

2-Methyl-octahydro-quinolin-4-one

  • MDL number MFCD02932745
  • Cas Number : 41854-83-5
  • Molecular Weight : 167.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-128
Density 1.0290/20
Country of Origin USA
Boiling point 108�orr
Product Link http://www.matrixscientific.com/010369.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010369.html
Availability details for
Quantity
  • Availability
  • Add to Cart