MAT047029229 Matrix Scientific

1-Isopropyl-1H-indole-3-carbaldehyde

  • MDL number MFCD02629696
  • Cas Number : 151409-84-6
  • Molecular Weight : 187.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010375.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010375.html
Availability details for
Quantity
  • Availability
  • Add to Cart