MAT047029230 Matrix Scientific

5-(1H-Benzoimidazol-2-ylsulfanyl)-furan-2-carbaldehyde

  • MDL number MFCD00442014
  • Cas Number : 39689-08-2
  • Molecular Weight : 244.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-163
Country of Origin USA
Product Link http://www.matrixscientific.com/010376.html

Safety Information

MSDS http://www.matrixscientific.com/010376.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart