MAT047029233 Matrix Scientific

5-Azepan-1-yl-furan-2-carbaldehyde

  • MDL number MFCD02629620
  • Cas Number : 436088-58-3
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010379.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010379.html
Availability details for
Quantity
  • Availability
  • Add to Cart